- Description:Spill kit, multipurpose, 1 piece
- Storage temperature:RT
- Packed:Fibre carton
- Pk:1 KIT
For safe, easy, effective spill treatment in the laboratory.
- Agents are formulated to rapidly clean up chemical spills and vapors
- Each bottle has a convenient dispensing cap for shaking or pouring
Caustics and acids are converted into nonhazardous compounds that meet EPA specifications for nonhazardous waste, simplifying cleanup and disposal. Solvent spills can be easily treated and transported with the absorbent in the solvent spill kit. This absorbent will also act as a vapor suppressant. The formaldehyde spill kit contains a two-part neutralisation and immobilisation system that effectively neutralises formaldehyde solution spills. The multipurpose kit can treat up to a 4 L solvent spill or a 2,5 L acid and caustic spill. The mercury spill kit controls mercury spills in minutes using a simple, two-step procedure. Also included is a mercury vapor suppressant absorbent which is ideally suited to penetrate inaccessible areas such as cracks and crevices.
SpillSolv® kits are available for acid, caustic, solvent, formaldehyde, or mercury spills. Also available is a multipurpose spill kit that contains spill response materials for a combination of spills.