VWR® Extractor™ Ethidium Bromide Waste Reduction System
Catalog # VWRMNP10448031
Supplier: VWR Products
Specifications
- Description:Ethidium bromide waste reduction system
- Pk:6
Specifications
About this item
One-step filtration polypropylene funnel device for the rapid removal of ethidium bromide from gel-staining solutions.
It contains an activated carbon matrix, which removes >99% of ethidium bromide from electrophoretic buffer quickly and easily.
The extractor funnel device fits most standard laboratory flasks and bottles (neck size 33 to 45 mm), and the unit includes a cap for storage between uses. The polypropylene housing is chemically resistant to organics. Pack includes glass fiber prefilters, which remove gel pieces and other debris to avoid premature clogging of the carbon filter.
Each device can decontaminate up to 10 litres of gel-staining solution. The decontaminated solution can be safely poured down the laboratory drain.