VWR® Dykem® High Temperature 44 Markers
VWRMNP253-44219
- Colour:White
- Description:High temperature resistant action Marker®, 843,3 to 898,9 °C, 54,43 g weight
- Pk:12
High temperature marker for use on glass, most metals or ceramic.
- Medium size fiber tip
- Pigmented and oil-based
- Pen cap clip fits on pocket or clipboard
- Marks become permanent and fused when heated
Full cure time: 1 min at 25 °C