Order Entry
Singapore
ContactUsLinkComponent
Petroleum spirit, 40…60 °C, Analytical reagent (AR), Rankem®
Flammable
Irritant
Health
Environment
Petroleum spirit, 40…60 °C, Analytical reagent (AR), Rankem®
Catalog # RANKP0072
Supplier:  RANKEM
CAS Number:  
Flammable
Irritant
Health
Environment
Petroleum spirit, 40…60 °C, Analytical reagent (AR), Rankem®
Catalog # RANKP0072
Supplier:  RANKEM
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, amber
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Color (APHA):
    ≤ 10
  • Weight Per ml (at 20 °C):
    0,640 - 0,655 g
  • Boiling Range (95%):
    40.0 - 60.0°C
  • Residue After Evaporation:
    ≤ 0,001%
  • Acidity:
    Passes test
  • Water (By KF):
    ≤ 0,02%
  • Substances Discoloured By H₂SO₄:
    Passes test
  • Copper (Cu):
    ≤ 0,0001%
  • Iron (Fe):
    ≤ 0,0001%
  • Lead (Pb):
    ≤ 0,0001%

Specification Test Results

About this item

 :
Boiling Pt:40…60 °C (1013 hPa)
Density:0,645…0.665g/cm³ (20 °C)
Flash Pt:–40 °C
EINECS:265-151-9
UN:1268
ADR:3,II
 :GHS02
 :GHS07
 :GHS08
 :GHS09
CAS Number:8032-32-4
MDL Number:MFCD00081849