Order Entry
Singapore
ContactUsLinkComponent
Toluene ≥99.00%, Laboratory Reagent sulphur-free, Rankem®
Flammable
Health
Irritant
Toluene ≥99.00%, Laboratory Reagent sulphur-free, Rankem®
Catalog # RANKT0150
Supplier:  RANKEM
CAS Number:  
Flammable
Health
Irritant
Toluene ≥99.00%, Laboratory Reagent sulphur-free, Rankem®
Catalog # RANKT0150
Supplier:  RANKEM
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, amber
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Assay (GC):
    Min. 99.00%
  • Weight Per ml (at 20 °C):
    0.864 --> 0.867 g
  • Refractive Index (at 20 °C):
    1.493 --> 1.497
  • Distillation Range:
    109.0 - 111.0°C
  • Non volatile matter:
    Max. 0.003%
  • Sulphur Compounds (As SO₄²⁻):
    ≤ 0.0005%
  • Thiophene Homologues:
    ≤ 0.0002%
  • Water:
    Max. 0.04%
  • Readily Carbonisable Substances:
    Passes test

Specification Test Results

About this item

 :
 :92.14 g/mol
Merck Index:13,09607
Boiling Pt:110,6 °C (1013 hPa)
Melting Pt:–95 °C
Density:0,867 g/cm³ (20 °C)
Flash Pt:4 °C
Storage Temperature:Ambient
EINECS:203-625-9
UN:1294
ADR:3,II
REACH:01-2119471310-51
 :GHS02
 :GHS08
 :GHS07
CAS Number:108-88-3
Formula:C₆H₅CH₃
MDL Number:MFCD00008512