



Formaldehyde solution 37% LR stabilised, Laboratory Reagent, Rankem®
Supplier: RANKEM
Certificates
About this item
CAS Number:50-00-0
:
Formula:CH₂O
MDL Number:MFCD00003274
:30.03 g/mol
Melting Pt:–15 °C
Density:1,08 g/cm³ (20 °C)
Flash Pt:60 °C (closed cup)
UN:1198
ADR:3,III
:Stabilised with methanol
:GHS02
:GHS05
:GHS06
:GHS08
Specification Test Results
- Guarantee Analysis:.
- Assay:36.50 --> 38.00 %
- Weight Per ml (at 20°C):1.084 --> 1.090 g
- Methanol (Stabilizer):10.00 --> 14.00 %
- Maximum Limits Of Impurities:Jun-03
- Acidity (As HCOOH):Max. 0.050 %
Specification Test Results
Related Information
Certificates
Search by Lot Numberspec_RANKF0080 spec_RANKF0081 spec_RANKF0090 spec_RANKF0100 spec_RANKF0107