

Chloroform ≥99.00% stabilised ACS, Analytical reagent (AR), Rankem®
Supplier: RANKEM
Certificates
About this item
CAS Number:67-66-3
:
Formula:CHCl₃
MDL Number:MFCD00000826
:119.38 g/mol
Merck Index:13,02160
Boiling Pt:61,7 °C (1013 hPa)
Melting Pt:–63 °C
Density:1,4832 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:1888
ADR:6.1,III
:Stabilised with ethanol 0,6%
EINECS:200-663-8
REACH:01-2119486657-20
:GHS06
:GHS08
Specification Test Results
- Colour (APHA):≤ 10
- Guarantee Analysis:.
- Assay(GC):99.00 --> 99.40%
- Assay (According To ACS) (GC) CHCl3:Min. 99.80%
- Alcohol (As Stabiliser):0.60 --> 1.00%
- Weight Per ml (at 20 °C):1.474 --> 1.481 g
- Identity (IR):Conforms
- Maximum Limits Of Impurities:.
- Residue After Evaporation:Max. 0.0005%
- Acidity (as HCl):Max. 0.0005%
- Water ( By KF):Max. 0.05%
- Acid And Chloride:Passes test
- Aldehydes And Ketones (As C3H6O):<=0.005%
- Free Chlorine (Cl):Passes test
- Substances Darkened By Sulphuric Acid:Passes test
- Suitability For Use With Dithizone:Passes test
- Dichloromethane (GC):Max. 0.10%
- Carbon Tetrachloride (GC):Max. 0.01%
- Trichloroethylene (GC):Max. 0.01%
- Tetrachloroethylene (GC):Max. 0.01%
- Chloride (Cl):<=0.00002%
- Copper(Cu):Max. 0.0001%
- Iron (Fe):Max. 0.0001%
- Lead ( Pb):Max. 0.0001%