VWR® 96-Well Deep Well Plate, Polypropylene
VWRU76210-522
- Sterile:+
- Colour:Clear
- Description:96 well deep well plate, polypropylene, conical bottom, round well shape
- Pk:50
- Well volume:0,8 ml
VWR® 96-well deep well plates are ideal for sample storage, sample re-suspension, cell culture, assay setup, diluting, and compound libraries. They are compatible with automated systems and are widely used in laboratories. Different sealing options are available, including cap seals or film seals, which can be applied independently to avoid cross-contamination.
- DNase-free, RNase-free, and non-pyrogenic
- Free from BSE and TSE
- Latex-free
- Sterilised by gamma irradiation to SAL 10⁻⁶ (ISO 11137)
- Three years shelf life following the month of production
The 96-well deep well storage plates feature round well shapes, and are made of medical grade, virgin polypropylene resins. The round well shape is designed for optimal sample recovery. Plates are manufactured in a class 100000 cleanroom environment to avoid any contaminations and guarantee high-quality. They are packed in plastic bags to avoid contamination.
: Manufactured under EN ISO 13485 and ISO 9001 quality management systems.
: Both sterile and non sterile formats are available.