



n-Pentane ≥99.9%, B&J Brand™ for HPLC, for gas chromatography, for spectrophotometry, Burdick & Jackson™
Catalog # BJAA312-4
Supplier: BURDICK JACKSON
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:16 L
- Seq:5254
Specifications
Specification Test Results
- Electron Capture GC:10 ng/L max.
- GC Analysis (n-Pentane):98 % min.
- GC Analysis (total C5 isomers):99.9 % min.
- Refractive Index (20ºC):1.3573 - 1.3579
- Residue:1 mg/L max.
- UV Cutoff:190 nm max.
- Water by Karl Fischer Titration:0.01 % max.
Specification Test Results
About this item
:
:72.15 g/mol
Merck Index:12,07255
Boiling Pt:36 °C (1,013.25 hPa)
Melting Pt:–129,7 °C
Density:0,626 g/cm³ (20 °C)
Flash Pt:7,8 %(V)
Storage Temperature:Ambient
EINECS:203-692-4
UN:1265
ADR:3,II
:GHS02
:GHS08
:GHS07
:GHS09
CAS Number:109-66-0
Formula:H₃C(CH₂)₃CH₃
MDL Number:MFCD00009498