Order Entry
Singapore
ContactUsLinkComponent
2-Propanol ≥99.80% AR ACS, Analytical reagent (AR), Rankem®
Flammable
Irritant
2-Propanol ≥99.80% AR ACS, Analytical reagent (AR), Rankem®
Catalog # RANKP0674
Supplier:  RANKEM
CAS Number:  
Flammable
Irritant
2-Propanol ≥99.80% AR ACS, Analytical reagent (AR), Rankem®
Catalog # RANKP0674
Supplier:  RANKEM
CAS Number:  

Specifications

  • Pack type:
    HDPE Jerry can
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Guarantee Analysis:
    .
  • Assay(GC):
    ≥ 99.80 %
  • Color (APHA):
    ≤ 10
  • Identity (IR):
    Conforms
  • Weight Per ml (at 20 °C):
    0.784 - 0.786 g
  • Solubility In Water:
    Passes test
  • Maximum Limits Of Impurities:
    .
  • Residue After Evaporation:
    ≤ 0.001 %
  • Water ( By KF):
    ≤ 0.10 %
  • Acidity:
    ≤ 0.0001 meq/g
  • Alkalinity:
    ≤ 0.0001 meq/g
  • Carbonyl Compounds (as HCHO):
    ≤ 0.002 %
  • Impurities Detected By Glc:
    ≤ 0.20 %
  • Methanol (GC):
    ≤ 0.10 %
  • Readily Carbonisable Substances:
    Passes test
  • Substances Reducing KMnO4(as O):
    <=0.0005%
  • Copper(Cu):
    ≤ 0.01 ppm
  • Iron (Fe):
    ≤ 0.0001 %
  • Lead ( Pb):
    ≤ 0.50 ppm

Specification Test Results

About this item

 :
 :60.1 g/mol
Boiling Pt:82 °C (1013 hPa)
Melting Pt:–89 °C
Density:0,786 g/cm³ (20 °C)
Flash Pt:12 °C
EINECS:200-661-7
UN:1219
ADR:3,II
 :GHS02
 :GHS07
CAS Number:67-63-0
Formula:(CH₃)₂CHOH
MDL Number:MFCD00011674