


Methyl red solution 0,1% (w/v) alcoholic solution USP Test Solution (TS)
Catalog # RICCR5045000-500A
Supplier: Ricca Chemical
CAS Number:
Specifications
- Pack type:Poly bottle
- Pk:500 mL
- Seq:2725
Specifications
Specification Test Results
- Appearance:Clear, dark red, alcoholic odor
- Suitability for Use:Pink (pH 6.2)
Specification Test Results
About this item
:
Boiling Pt:~ 77 °C
Density:0,8000 g/cm³
Flash Pt:~15,5 °C
ADR:3,II
:269.3 g/mol
UN:1987
:GHS08
:GHS07
:GHS02
Storage Temperature:Ambient
CAS Number:493-52-7
Formula:C₁₅H₁₅N₃O₂
MDL Number:MFCD00002425