$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Singapore
Orders Link
ContactUsLinkComponent
Enzymes
Trypsin
Trypsin
Glass bottle for solids, 100 mg
EDQMT2600000
:
EDQM
Trypsin
EDQMT2600000
:
EDQM
:
Trypsin
Glass bottle for solids, 100 mg
:
$1,940.00
1 * 100 mg
-
+
Pack type:
Glass bottle for solids
Pk:
100 mg
Cat. No.:
EDQMT2600000
Storage conditions:
Refrigerator
MDL No.:
MFCD00082094
Enzyme name:
Trypsin
CAS No.:
9002-07-7