Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Starch
Starch 1% (w/v) in aqueous solution for iodometric titrations mercury-free
Starch 1% (w/v) in aqueous solution for iodometric titrations mercury-free
Starch 1% (w/v) in aqueous solution for iodometric titrations mercury-free
RICC8050-32
:
Ricca Chemical
:
Starch 1% (w/v) in aqueous solution for iodometric titrations mercury-free
RICC8050-32
:
Ricca Chemical
:
Pack type:
Glass bottle
Pk:
1 L
Seq:
5037
:
:
162.14 g/mol
:
Ambient
:
232-679-6
:
9005-25-8
:
(C₆H₁₀O₅)n
:
MFCD00082026