



Petroleum spirit, 20…70 °C ≥99.0% (by GC) (C5 & C6 Isomers), B&J Brand™ for gas chromatography, for pesticide residue analysis, Burdick & Jackson™
Catalog # BJAA317-4
Supplier: BURDICK JACKSON
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:16 L
- Seq:5254
Specifications
Specification Test Results
- Electron Capture GC:10 ng/L max.
- GC Analysis (total C5 and C6 isomers):99 % min.
- Refractive Index (20ºC):1.36 - 1.37
- Residue:1 mg/L max.
- Water by Karl Fischer Titration:0.01 % max.
Specification Test Results
About this item
:
Boiling Pt:35…60 °C
Melting Pt:–73 °C
Density:0,64 g/cm³ (20 °C)
Flash Pt:5,9 %(V)
Storage Temperature:Ambient
EINECS:309-852-0
UN:1268
ADR:3,II
:GHS02
:GHS08
:GHS07
:GHS09
CAS Number:64742-49-0
:64742-82-1
MDL Number:MFCD00081849