$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Singapore
ContactUsLinkComponent
Methyl red
Methyl red 0,1% (w/v) in aqueous solution indicator
Methyl red 0,1% (w/v) in aqueous solution indicator
RICC5060-32
:
Ricca Chemical
:
Methyl red 0,1% (w/v) in aqueous solution indicator
RICC5060-32
:
Ricca Chemical
:
Pk:
1
Seq:
0001
:
:
269.3 g/mol
:
Ambient
:
212-682-9
:
493-52-7
:
C₁₅H₁₅N₃O₂
:
MFCD00002425