Order Entry
Singapore
Orders LinkContactUsLinkComponent
Colchicine

Colchicine

Toxic
Health
Supplier:   PanReac AppliChem
Colchicine
Supplier:  PanReac AppliChem
Certificates

2 Options Available Below

Product Details & Documents
Synonyms:  7α-H-Colchicine, (S)-(-)-Colchicine
  • CAS Number:64-86-8
  • Formula:C₂₂H₂₅NO₆
  • MDL Number:MFCD00078484
  • Molecular Weight:399.44 g/mol
  • Merck Index:12,02536
  • Melting Pt:154…156 °C
  • Density:1,32 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-598-5
  • UN:1544
  • ADR:6.1,I

Documents

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - $1,270.00
    $1,270.00
    1 * 1 g
       $1,270.00

  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - $4,990.00
    $4,990.00
    1 * 5 g
       $4,990.00