WHEATON® Boston Round Bottles, Clear, Narrow Mouth
Catalog # WHEA14235-666
Supplier: DWK Life Sciences
Specifications
- Cap Size:24-400
- Capacity:250 ml
- Description:Clear Boston round bottles with poly-vinyl lined white polypropylene cap, Ø×H: 60×136 mm
- Pk:108
Specifications
About this item
Wheaton Boston round bottles are manufactured from clear soda lime glass that conforms to USP Type III requirements.
These narrow mouth bottles facilitate pouring and storing of liquids. Each case of bottles is packaged in convenience packs with caps attached to maintain cleanliness. Partitioned carton protects bottles from scratches during shipping, and they become useful for storing samples. Each case of bottles also includes a hazardous communication label and tamper evident seal if labeling and/or sealing is required.
Ordering information: Available with caps attached or in bulk packs without caps. When included, caps come attached to bottles. Extra screw caps can be purchased separately.