Water, B&J Brand™ LC-MS for LC-MS, for HPLC, Burdick & Jackson™
:7732-18-5
:
:H₂O
:MFCD00011332
:18.02 g/mol
:13,10098
:100 °C
:0 °C
:0,9971 g/cm³ (25 °C)
:Ambient
:231-791-2
- Fluorescence (as quinine base):0.3 ppb max.
- HPLC Gradient Elution:Passes test
- LC-MS Negative Mode (as 4-nitrophenol):50 ppb max.
- LC-MS Positive Mode (as caffeine):100 ppb max.
- LC-MS Positive Mode (as reserpine):50 ppb max.
- Refractive Index (20ºC):1.3325 - 1.3335
- Residue:1 mg/L max.
- Trace Metals:Passes test
- Ultraviolet Absorbance Test:Passes test