Flasks, flat bottom, narrow or wide neck, with rim
SCOT21373136
- Capacity:250 ml
- Description:Flask flat bottom, wide/narrow neck, with beaded rim
- Environmentally Preferable:
- Pk:10
DURAN®, borosilicate glass 3.3, clear.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes