



n-Heptane ≥99% for HPLC
Catalog # RANKH2043
Supplier: RANKEM
CAS Number:
Specifications
- Pack type:Amber colored glass bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Color (APHA):≤ 10
- Guarantee Analysis:.
- Assay(GC):Min. 99.00 %
- Weight Per ml (at 20°C):0.683 --> 0.684 g
- Identity (IR):Conforms
- Maximum Limits Of Impurities:.
- Residue After Evaporation:Max. 0.0005 %
- Titrable Acid:Max. 0.0002 meq/g
- Titrable Base:Max. 0.0002 meq/g
- Water ( By KF):Max. 0.02 %
- UV Transmittance:.
- Wavelength:.
- 210 nm:Min. 50 %
- 220 nm:Min. 80 %
- 230 nm:Min. 90 %
- From 250 nm:Min. 98 %
Specification Test Results
About this item
:
:100.20 g/mol
Boiling Pt:98,4 °C (1013 hPa)
Melting Pt:–90,6 °C
Density:0,6838 g/cm³ (20 °C)
Flash Pt:−4 °C
EINECS:205-563-8
UN:1206
ADR:3,II
:GHS02
:GHS07
:GHS08
:GHS09
CAS Number:142-82-5
Formula:H₃C(CH₂)₅CH₃
MDL Number:MFCD00009544