Order Entry
Singapore
Orders LinkContactUsLinkComponent
Potassium Chloride Reference Solutions A - D
Potassium Chloride Reference Solutions A - D
Potassium Chloride Reference Solutions A - D
Potassium Chloride Reference Solutions A - D
Potassium Chloride Reference Solutions A - D
Product Family Options
  • Description
    Conductivity
    Reference temperature
    Contents
    Pk
  • Potassium chloride reference solution A
    111,342 mS/cm (111,342 µmho/cm) at 25 °C
    25 °C
    1 Molar
    1 L
    1 * 1 L
    1 * 1 L - $860.00
    $860.00
    1 * 1 L
       $860.00
  • Potassium chloride reference solution C
    1408,8 µS/cm (1408.8 µmho/cm) at 25 °C
    25 °C
    0,01 Molar
    1 L
    1 * 1 L
    1 * 1 L - $806.00
    $806.00
    1 * 1 L
       $806.00