GHS Labels 'Corrosive Picto'
Catalog # BRDY121198
Supplier: Brady
Specifications
- Description:GHS picto labels
- Dimensions:38,1×38,1 mm
- Material:Polyester
- Packed:20 labels/card
- Pk:1
- Legend:Corrosive Picto
Specifications
About this item
Pre-printed GHS pictogram die cut labels on a card.
- Colour: Black/red on white
- Corrosive picto type of label