White Marker Pen
Catalog # VWRMNP4TI-0394
Supplier: VWR Products
Specifications
- Colour:White
- Description:White marker pen
- Pk:1
Specifications
About this item
This white marker pen is ideal for labeling Optically Clear Tissue Culture Plates, FrameStar® plates, 8 Well PCR Tube Strips with PC Frame, and other colored microplates or PCR plates.
- White ink ensures clear labeling details
- Ideal for labeling colored microplates or PCR plates
- Ink is resistant to aqueous or organic solvents
The resulting waterproof ink cannot be removed using aqueous or organic solvents.
Packaging: One pen per case.