THM reagent 3
Catalog # HACH2756699
Supplier: Hach
CAS Number:
Specifications
- Chemical method:THM Plus™ Hach Method 10132
- Pk:100
- Type:Powder pillows
- Tests per kit:50
Specifications
About this item
Screening tool to optimise treatment processes, to locate areas of high THM levels in distribution systems and to determine levels of trihalomethanes in drinking water samples.
- For determination of Total Trihalomethanes by the THM Plus™ Method
- Hach Method 10132