Order Entry
Singapore
ContactUsLinkComponent
THM reagent 1
THM reagent 1
Catalog # HACH2753929
Supplier:  Hach
CAS Number:  
undefined
THM reagent 1
Catalog # HACH2753929
Supplier:  Hach
CAS Number:  
Specifications
  • Chemical method:
    THM Plus™ Hach Method 10132
  • Pack type:
    Dropping bottle, self-contained
  • Pk:
    1 KIT
  • Type:
    Solution
  • Tests per kit:
    50
Specifications
About this item
Screening tool to optimise treatment processes, to locate areas of high THM levels in distribution systems and to determine levels of trihalomethanes in drinking water samples.
  • For determination of Total Trihalomethanes by the THM Plus™ Method
  • Hach Method 10132
Caution: Corrosive, Caution