Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
di-Potassium hydrogen phosphate
di-Potassium hydrogen phosphate ACS
di-Potassium hydrogen phosphate ACS
di-Potassium hydrogen phosphate ACS
Catalog # HAWC34263
Supplier:
HAWKINS CHEMICAL
CAS Number:
Expand image
di-Potassium hydrogen phosphate ACS
Catalog # HAWC34263
Supplier:
HAWKINS CHEMICAL
CAS Number:
Specifications
Pk:
500 g
Seq:
3645
Specifications
About this item
Product Details
:
:
174.18 g/mol
CAS Number:
7758-11-4
Formula:
K₂HPO₄
MDL Number:
MFCD00011383
Product Details