

Acetonitrile ≥99.9%, tested NMR, gradient grade for HPLC, Rankem®
RANKA2096
: RANKEM
:
- Pack type:Amber colored glass bottle
- Pk:2,5 L
- Seq:5096
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussion
- Guarantee Analysis:.
- Assay(GC):Min. 99.90 %
- Weight Per ml (at 20°C):0.782 -- 0.783 g
- Maximum Limits Of Impurities:.
- Acidity:Max. 0.0002 meq/g
- Alkalinity:Max. 0.0002 meq/g
- Non volatile matter:Max. 0.0004 %
- Water ( By KF):Max. 0.03 %
- UV Transmittance:.
- Wavelength:.
- 195 nm:Min. 75 %
- 200 nm:Min. 93 %
- 210 nm:Min. 96 %
- 220 nm:Min. 97 %
- from 230 nm:Min. 98 %
- Gradient Performance:.
- at 254 nm:≤ 1 mAU
- at 210 nm:≤ 5 mAU
- NMR Performance:Passes test
:
:41.05 g/mol
:13,00071
:81,6 °C (1013 hPa)
:–45,7 °C
:0,782 g/cm³ (20 °C)
:2 °C (closed cup)
:Ambient
:200-835-2
:1648
:3,II
:Filtered through a 0.2 µm filter.
:01-2119471307-38
:GHS02
:GHS07
:75-05-8
:H₃CCN
:MFCD00001878