Eppendorf® Safe-Lock 1,5 ml microtubes ensure secure sample storage. With a patented Safe-Lock design, these microtubes prevent accidental opening, safeguarding samples. Ideal for diverse applications, they offer reliability, durability, and ease of use in a 1,5 ml capacity.
- Hinged Safe-Lock lid prevents inadvertent lid opening during incubation and storage
- g-Safe® technology, depending on the Safe-Lock Tube variant, ensures outstanding centrifugation stability up to 30,000×g
- g-Safe® technology prevents sample loss due to tube breakage, and provides additional safety when handling hazardous samples
- Polypropylene construction offers robust resistance to chemicals, mechanical stress, and extreme temperatures
- Precision lid sealing minimises evaporation rates during extended storage periods
- Generously sized frosted lid and side surface facilitate easy labelling
- Assured functionality in a temperature range from −86 to 100 °C
- Autoclavable when open (121 °C, 20 min) for reliable sterilisation
- Available in lot-certified PCR clean, Eppendorf Forensic DNA Grade, and Biopur® quality for tested purity
Eppendorf® Safe-Lock 1,5 ml microtubes exemplify secure sample storage with their patented Safe-Lock design, preventing accidental openings and preserving sample integrity. Versatile across laboratory applications, the durable construction and 1,5 ml capacity cater to varied sample volumes.
Clear graduations and a smooth surface enhance user-friendliness, ensuring a seamless and reliable laboratory experience. Elevate scientific endeavors with the trusted quality of Eppendorf® Safe-Lock Tubes.
Manufactured without the use of slip agents, plasticisers, and biocides - substances that have been shown to leach from plastic consumables into the sample and negatively affect bio-assay results.
- Catalog No:
- EPPE0030123.328
- EPPE0030120.086
- EPPE0030123.611
- EPPE0030121.589
- EPPE0030120.191
- EPPE0030120.167
- EPPE0030120.175
- EPPE0030120.159
- EPPE0030121.694
- Autoclavable:
- When open 121 °C, 20 min
- When open 121 °C, 20 min
- When open 121 °C, 20 min
- When open 121 °C, 20 min
- When open 121 °C, 20 min
- When open 121 °C, 20 min
- When open 121 °C, 20 min
- When open 121 °C, 20 min
- When open 121 °C, 20 min
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Operating temperature range:
- −86...100 °C
- −86...100 °C
- −86...100 °C
- −86...100 °C
- −86...100 °C
- −86...100 °C
- −86...100 °C
- −86...100 °C
- −86...100 °C
- Volume:
- 1,5 ml
- 1,5 ml
- 1,5 ml
- 1,5 ml
- 1,5 ml
- 1,5 ml
- 1,5 ml
- 1,5 ml
- 1,5 ml
- Environmental certification:
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001