


Acetonitrile : Water (50:50)% (v/v), LabReady™, Solvent Blend for HPLC, Burdick & Jackson™
Catalog # BJAA435-4
Supplier: BURDICK JACKSON
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:16 L
- Seq:5254
Specifications
Specification Test Results
- Water (v/v):49.0 - 51.0%
- Acetonitrile (v/v):49.0 - 51.0%
- UV Cutoff:186 nm
- Residue:< 1 mg/L
- Color (APHA):< 10
- HPLC Gradient @ 220 nm:< 0.005 AU
- HPLC Gradient @ 254 nm:< 0.001 AU
Specification Test Results
About this item
:
Boiling Pt:82 °C (solvent)
Density:0,7857 g/cm³ (20 °C) (pure substance)
Flash Pt:3 °C
Storage Temperature:Ambient
EINECS:200-835-2
UN:1648
ADR:3,II
:GHS08
:GHS07
:GHS02
:75-05-8