Order Entry
Singapore
ContactUsLinkComponent
Dichloromethane ≥99.9% stabilised ACS, meets analytical specification of USP for HPLC, for spectrophotometry
Irritant
Health
Dichloromethane ≥99.9% stabilised ACS, meets analytical specification of USP for HPLC, for spectrophotometry
Catalog # BJAAAH300-4
Supplier:  BURDICK JACKSON
CAS Number:  
undefined
Irritant
Health
Dichloromethane ≥99.9% stabilised ACS, meets analytical specification of USP for HPLC, for spectrophotometry
Catalog # BJAAAH300-4
Supplier:  BURDICK JACKSON
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    16 L
  • Seq:
    5254

Specifications

Specification Test Results

  • Appearance:
    Passes test
  • Color (APHA):
    10 max.
  • Free Halogens:
    Passes test
  • GC Analysis:
    99.9 % min.
  • Refractive Index (20ºC):
    1.4236 - 1.4246
  • Residue:
    3 mg/L max.
  • Titrable Acid:
    0.3 µeq/g max.
  • UV Cutoff:
    233 nm max.
  • Water by Karl Fisher Titration:
    0.02 % max.

Specification Test Results

About this item

 :
 :84.93 g/mol
Merck Index:12,06140
Boiling Pt:40 °C
Melting Pt:–95 °C
Density:1,33 g/cm³
Flash Pt: Does not flash
EINECS:200-838-9
UN:1593
ADR:6.1,III
 :Stabilised with cyclohexene
 :GHS07
 :GHS08
Storage Temperature:Ambient
CAS Number:75-09-2
Formula:CH₂Cl₂
MDL Number:MFCD00000881