Order Entry
Singapore
ContactUsLinkComponent
Hexane (mixture of isomers) ≥99.9% (n-Hexane > 95%), GC2™ for gas chromatography, for pesticide residue analysis, Burdick & Jackson™
Irritant
Health
Flammable
Environment
Hexane (mixture of isomers) ≥99.9% (n-Hexane > 95%), GC2™ for gas chromatography, for pesticide residue analysis, Burdick & Jackson™
Catalog # BJAAGC215-4
Supplier:  BURDICK JACKSON
CAS Number:  
undefined
Irritant
Health
Flammable
Environment
Hexane (mixture of isomers) ≥99.9% (n-Hexane > 95%), GC2™ for gas chromatography, for pesticide residue analysis, Burdick & Jackson™
Catalog # BJAAGC215-4
Supplier:  BURDICK JACKSON
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    16 L
  • Seq:
    5254

Specifications

Specification Test Results

  • Electron Capture GC (as lindane):
    2 ppt max.
  • Flame Ionization GC (as nonane):
    1 ppb max.
  • GC Analysis (n-Hexane):
    95 % min.
  • GC Analysis (total C6 isomers):
    99.9 % min.
  • Residue:
    1 mg/L max.
  • Water by Karl Fischer Titration:
    0.01 % max.

Specification Test Results

About this item

 :
 :86.18 g/mol
Merck Index:13,04712
Boiling Pt:7,7 %(V)
Melting Pt:–95 °C
Density:0,659…0.673 g/cm³ (20 °C)
Flash Pt:–26 °C
Storage Temperature:Ambient
EINECS:203-777-6
UN:1208
ADR:3,II
 :GHS07
 :GHS08
 :GHS02
 :GHS09
CAS Number:110-54-3
 :92112-69-1
Formula:H₃C(CH₂)₄CH₃
MDL Number:MFCD02179311