Dicyclohexyl sulphide, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:2550-40-5
:
Formula:C₁₂H₂₂S₂
MDL Number:MFCD00013759
:230.44 g/mol
Boiling Pt:73…75 °C (0,04 mmHg)
Density:1,0457 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ300
EINECS:219-851-6