
Diethylene glycol monomethyl ether
Catalog # EDQMY0000217
Supplier: EDQM
CAS Number:
Specifications
- Pk:1
- Seq:0001
Specifications
About this item
:
:120.15 g/mol
Merck Index:12,06116
Boiling Pt:193 °C (1013 hPa)
Melting Pt:–65 °C
Density:1,025 g/cm³ (20 °C)
Flash Pt:91 °C
Storage Temperature:Ambient
EINECS:203-906-6
:GHS08
CAS Number:111-77-3
Formula:C₅H₁₂O₃
MDL Number:MFCD00002871