VWR® Double Edge Blades, 10 Blade Dispenser
Catalog # VWRMAGBL-6007-0000
Supplier: VWR Products
Specifications
- Description:Double edge blades, oiled, 10 blade dispenser, 500 dispensers/case
- Material:Stainless steel
- Thickness:0.10 mm
- Pk:10
Specifications
About this item
Double edge blades feature a 3-Facet edge for exacting, clean cuts. Safety blade dispenser holds 10 blades.
- Blade edge with MicroCoat technology enhances durability and sharpness
- Facet edge for increased sharpness
The MicroCoat enrichment technology reduces cut force and adds a layer of protection for each blade helping the blade maintain a sharp effective edge over multiple uses.
Caution: Blades are extremely sharp. Wear eye protection. Handle and use with care. Bending blades may break them. Keep out of children's reach. Dispose of safely.