Di-tert-butyl peroxide, Sigma-Aldrich®
Catalog # SAFA168521-250ML
Supplier: Merck
CAS Number:
Specifications
- Pk:250 mL
- Seq:2590
Specifications
About this item
:
:146.23 g/mol
Merck Index:12,03515
Boiling Pt:109 °C (1013 hPa)
Melting Pt:< –25 °C
Density:0,798 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Flash Pt:12 °C
EINECS:203-733-6
:GHS02
CAS Number:110-05-4
Formula:C₈H₁₈O₂
MDL Number:MFCD00008803