Gemfibrozil
196 µG
Catalog # EDQMY0000514
Supplier: EDQM
Jump to:
Product Details & Documents
Synonyms: 5-(2,5-Dimethylphenoxy)-2,2-dimethylpentanoic acid, 2,2-Dimethyl-5-(2,5-dimethylphenoxy)pentanoic acid, CI-719, 2,2-Dimethyl-5-(2,5-dimethylphenoxy)valeric acid
- CAS Number:25812-30-0
- Formula:C₁₅H₂₂O₃
- MDL Number:MFCD00079335
- Molecular Weight:250.34 g/mol
- Merck Index:12,04394
- Boiling Pt:395 °C (1013 hPa)
- Melting Pt:59…61 °C
- EINECS:247-280-2
Specifications
Specifications
- Pk:196 µG



