


Toluene ≥99.8%, Purified Plus™ ACS for organic synthesis, for preparative liquid chromatography, Burdick & Jackson™
Supplier: BURDICK JACKSON
Certificates
About this item
CAS Number:108-88-3
:
Formula:C₆H₅CH₃
MDL Number:MFCD00008512
:92.14 g/mol
Merck Index:13,09607
Boiling Pt:7,1 %(V)
Melting Pt:–95 °C
Density:0,867 g/cm³ (20 °C)
Flash Pt:4 °C
Storage Temperature:Ambient
EINECS:203-625-9
UN:1294
ADR:3,II
REACH:05-2114615130-69
:GHS07
:GHS08
:GHS02
Specification Test Results
- Assay by GC Analysis:99.8 % min.
- Color (APHA):10 max.
- Refractive Index (20ºC):1.4957 - 1.4977
- Residue after Evaporation:5 mg/L max.
- Substances Darkened by Sulfuric Acid:Passes test
- Sulfur Compounds (as S):0.003 % max.
- Water by Karl Fischer Titration:0.03 % max.