

Dichloromethane ≥99.9% stabilised, Purified Plus™ ACS for organic synthesis, for preparative liquid chromatography, Burdick & Jackson™
Supplier: BURDICK JACKSON
Certificates
About this item
CAS Number:75-09-2
:
Formula:CH₂Cl₂
MDL Number:MFCD00000881
:84.93 g/mol
Merck Index:12,06140
Boiling Pt:40 °C
Melting Pt:–95 °C
Density:1,33 g/cm³
Flash Pt: Does not flash
UN:1593
ADR:6.1,III
:Stabilised with cyclohexene
EINECS:200-838-9
:GHS07
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Appearance:Passes test
- Assay by GC Analysis:99.9 % min.
- Color (APHA):10 max.
- Free Halogens:Passes test
- Refractive Index (20ºC):1.4231 - 1.4251
- Residue after Evaporation:5 mg/L max.
- Titrable Acid:0.3 µeq/g max.
- Water by Karl Fisher Titration:0.02 % max.