Methanol ≥99.9%, Purified Plus™ ACS for organic synthesis, for preparative liquid chromatography, Burdick & Jackson™
:67-56-1
:
:CH₃OH
:MFCD00004595
:32.04 g/mol
:13,05984
:64,7 °C
:–98 °C
:0,792 g/cm³ (20 °C)
:11 °C
:Ambient
:200-659-6
:1230
:3,II
:01-2119433307-44
:GHS02
:GHS08
- Assay by GC Analysis:99.9 % min.
- Water by Karl Fischer Titration:0.1 % max.
- Carbonyl Compounds:0.001 % max.
- Refractive Index (20ºC):1.3274 - 1.3294
- Residue after Evaporation:5 mg/L max.
- Solubility in Water:Passes test
- Substances Darkened by Sulfuric Acid:Passes test
- Substances Reducing Permanganate:Passes test
- Titrable Acid:0.3 µeq/g max.
- Titrable Base:0.2 µeq/g max.