

Chloroform ≥99.9% stabilised, Purified Plus™ ACS for organic synthesis, for preparative liquid chromatography, Burdick & Jackson™
Supplier: BURDICK JACKSON
Certificates
About this item
CAS Number:67-66-3
:
Formula:CHCl₃
MDL Number:MFCD00000826
:119.38 g/mol
Merck Index:13,02160
Boiling Pt:62 °C
Melting Pt:–64 °C
Density:1,49 g/cm3
Storage Temperature:Ambient
UN:1888
ADR:6.1,III
:Stabilised with amylene
EINECS:200-663-8
REACH:05-2114614906-45
:GHS07
:GHS08
Specification Test Results
- Acetone and Aldehyde:Passes test
- Acid and Chloride:Passes test
- Assay by GC Analysis:99.9 % min.
- Color (APHA):10 max.
- Free Chlorine:Passes test
- Lead:0.05 ppm max.
- Refractive Index (20ºC):1.4439 - 1.4469
- Residue after Evaporation:5 mg/L max.
- Water by Karl Fischer Titration:0.05 % max.