Acetonitrile ≥99.8%, Purified Plus™ ACS for organic synthesis, for preparative liquid chromatography, Burdick & Jackson™
:75-05-8
:
:H₃CCN
:MFCD00001878
:41.05 g/mol
:13,00071
:82 °C(1,013 hPa)
:–46 °C
:0,7822 g/cm³ (20 °C)
:6 °C
:Ambient
:200-835-2
:1648
:3,II
:01-2119471307-38
:GHS07
:GHS08
:GHS02
- Appearance:Passes test
- Assay by GC Analysis:99.9 % min.
- Color (APHA):10 max.
- Refractive Index (20ºC):1.3430 - 1.3450
- Residue after Evaporation:5 mg/L max.
- Titrable Acid:8 µeq/g max.
- Titrable Base:0.6 µeq/g max.
- Water by Karl Fischer Titration:0.1 % max.