

2-Propanol ≥99.9%, B&J Brand™ for HPLC, for pesticide residue analysis, for spectrophotometry, Burdick & Jackson™
Supplier: BURDICK JACKSON
Certificates
About this item
CAS Number:67-63-0
:
Formula:(CH₃)₂CHOH
MDL Number:MFCD00011674
:60.1 g/mol
Merck Index:13,05228
Boiling Pt:82,3 °C
Melting Pt:–88 °C
Density:0,785 g/cm³ (20 °C)
Flash Pt:12 °C
Storage Temperature:Ambient
EINECS:200-661-7
UN:1219
ADR:3,II
REACH:01-2119457558-25
:GHS02
:GHS07
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default
Specifications
Specification Test Results
- Electron Capture GC:2 µg/L max.
- Water by Karl Fischer Titration:0.06 % max.
- GC Analysis:99.9 % min.
- Refractive Index (20ºC):1.3766 - 1.3778
- Residue:1 mg/L max.
- UV Absorbance @ 225 nm:0.16 AU max.
- UV Absorbance @ 250 nm:0.02 AU max.
- UV Absorbance @ 300 nm:0.005 AU max.
- UV Absorbance @ 400 nm:0.005 AU max.
- UV Cutoff:205 nm max.