N,N-Dimethylformamide ≥99.9%, B&J Brand™ for HPLC, for gas chromatography, for spectrophotometry, Burdick & Jackson™
Supplier: BURDICK JACKSON
Certificates
About this item
CAS Number:68-12-2
:
Formula:HCON(CH₃)₂
MDL Number:MFCD00003284
:73.09 g/mol
Merck Index:13,03269
Boiling Pt:153 °C
Melting Pt:–60,4 °C
Density:0,949 g/cm³ (20 °C)
Flash Pt:58 °C
Storage Temperature:Ambient
EINECS:200-679-5
UN:2265
ADR:3,III
REACH:01-2119475605-32
:GHS02
:GHS08
:GHS07
Specification Test Results
- GC Analysis:99.9 % min.
- GC Impurities Eluting Before Solvent:0.5 mg/L max.
- UV Absorbance @ 275 nm:0.3 AU max.
- UV Absorbance @ 300 nm:0.05 AU max.
- UV Absorbance @ 350 nm:0.005 AU max.
- UV Absorbance @ 400 nm:0.005 AU max.
- UV Cutoff:268 nm max.
- Water by Karl Fischer Titration:0.03 % max.