

2-Propanol ≥99.5% ACS for organic synthesis, for preparative liquid chromatography, general laboratory use
BJAALP323-4
:
- Pack type:Glass bottle
- Pk:16 L
- Seq:5254
- Appearance:Passes test
- Assay by GC Analysis:99.5 % min.
- Carbonyl Compounds:0.002 % max.
- Color (APHA):10 max.
- Residue after Evaporation:0.001 % max.
- Solubility in Water:Passes test
- Titrable Acid or Base:0.1 µeq/g max.
- Water by Karl Fishcer Titration:0.2 % max.
:
:60.1 g/mol
:13,05228
:12,0 %(V)
:–88 °C
:0,785 g/cm³ (20 °C)
:12 °C
:Ambient
:200-661-7
:1219
:3,II
:01-2119457558-25
:GHS07
:GHS02
:67-63-0
:(CH₃)₂CHOH
:MFCD00011674