

Dichloromethane ≥99.5% stabilised ACS for organic synthesis, for preparative liquid chromatography, general laboratory use
:75-09-2
:
:CH₂Cl₂
:MFCD00000881
:84.93 g/mol
:12,06140
:40 °C
:–95 °C
:1,33 g/cm³
: Does not flash
:1593
:6.1,III
:Stabilised with cyclohexene
:200-838-9
:GHS07
:GHS08
:Ambient
- Appearance:Passes test
- Assay by GC Analysis:99.5 % min.
- Color (APHA):10 max.
- Free Halogens:Passes test
- Residue after Evaporation:0.002 % max.
- Titrable Acid:0.3 µeq/g max.
- Water by Karl Fischer Titration:0.02 % max.