Methanol ≥99.8% ACS for organic synthesis, for preparative liquid chromatography, general laboratory use
Supplier: BURDICK JACKSON
Certificates
About this item
CAS Number:67-56-1
:
Formula:CH₃OH
MDL Number:MFCD00004595
:32.04 g/mol
Merck Index:13,05984
Boiling Pt:64,7 °C
Melting Pt:–98 °C
Density:0,792 g/cm³ (20 °C)
Flash Pt:11 °C
Storage Temperature:Ambient
EINECS:200-659-6
UN:1230
ADR:3,II
REACH:01-2119433307-44
:GHS02
:GHS08
Specification Test Results
- Assay by GC Analysis:99.8% min.
- Water by Karl Fischer Titration:0.1 % max.
- Carbonyl Compounds:0.001% max.
- Color (APHA):10 max.
- Residue After Evaporation:0.001% max.
- Solubility in Water:Passes test
- Substances Darkened by Sulfuric Acid:Passes test
- Substances Reducing Permanganate:Passes test
- Titrable Acid:0.3 µeq/g max.
- Titrable Base:0.2 µeq/g max.