


Acetonitrile ≥99.9%, B&J Brand™ LC-MS for LC-MS, for HPLC, Burdick & Jackson™
Supplier: BURDICK JACKSON
Certificates
About this item
CAS Number:75-05-8
:
Formula:H₃CCN
MDL Number:MFCD00001878
:41.05 g/mol
Merck Index:13,00071
Boiling Pt:82 °C (1,013 hPa)
Melting Pt:–46 °C
Density:0,7822 g/cm³ (20 °C)
Flash Pt:6 °C
Storage Temperature:Ambient
EINECS:200-835-2
UN:1648
ADR:3,II
REACH:01-2119471307-38
:GHS07
:GHS08
:GHS02
Specification Test Results
- GC Analysis:99.9 % min.
- Water by Karl Fischer Titration:0.01 % max.
- HPLC Gradient Elution:Passes test
- LC-MS Negative Mode (as 4-nitrophenol):50 ppb max.
- LC-MS Positive Mode (as caffeine):100 ppb max.
- LC-MS Positive Mode (as reserpine):50 ppb max.
- Refractive Index (20ºC):1.3434 - 1.3446
- Residue:1 mg/L max.
- Trace Metals:Passes test
- UV Cutoff:190 nm max.