Methanol ≥99.9%, GC2™ for gas chromatography, for pesticide residue analysis, Burdick & Jackson™
Catalog # BJAAGC230-4
Supplier: BURDICK JACKSON
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:16 L
- Seq:5254
Specifications
Specification Test Results
- Electron Capture GC (as lindane):2 ppt max.
- Flame Ionization GC (as nonane):1 ppb max.
- GC Analysis:99.9 % min.
- Refractive Index (20ºC):1.3280 - 1.3288
- Residue:1 mg/L max.
- Water by Karl Fischer Titration:0.1 % max.
Specification Test Results
About this item
:
:32.04 g/mol
Merck Index:13,05984
Boiling Pt:64,7 °C
Melting Pt:–98 °C
Density:0,792 g/cm³ (20 °C)
Flash Pt:11 °C
Storage Temperature:Ambient
EINECS:200-659-6
UN:1230
ADR:3,II
REACH:01-2119433307-44
:GHS02
:GHS08
CAS Number:67-56-1
Formula:CH₃OH
MDL Number:MFCD00004595