


Toluene, anhydrous ≥99.9% for applications requiring low water content, Burdick & Jackson™
Supplier: BURDICK JACKSON
Certificates
About this item
CAS Number:108-88-3
:
Formula:C₆H₅CH₃
MDL Number:MFCD00008512
:92.14 g/mol
Merck Index:13,09607
Boiling Pt:110,6 °C
Melting Pt:–95 °C
Density:0,867 g/cm3
Flash Pt:4 °C
Storage Temperature:Ambient
EINECS:203-625-9
UN:1294
ADR:3,II
REACH:05-2114615130-69
:GHS02
:GHS08
:GHS07
Specification Test Results
- Appearance:Passes test
- Color (APHA):10 max.
- GC Analysis:99.8 % min.
- Residue:3 mg/L max.
- Water by Karl Fischer Titration:50 ppm max.