

Dichloromethane, anhydrous ≥99.9% stabilised for applications requiring low water content, Burdick & Jackson™
Supplier: BURDICK JACKSON
Certificates
About this item
CAS Number:75-09-2
:
Formula:CH₂Cl₂
MDL Number:MFCD00000881
:84.93 g/mol
Merck Index:12,06140
Boiling Pt:40 °C
Melting Pt:–95 °C
Density:1,33 g/cm3
UN:1593
ADR:6.1,III
:Stabilised with amylene
EINECS:200-838-9
:GHS07
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Acidity (as HCl):1 mg/L max.
- Chloride:10 mg/L max.
- Color (APHA):10 max.
- GC Analysis:99.9 % min.
- Residue:3 mg/L max.
- Water by Karl Fischer Titration:50 ppm max.